Would you like to log in again?
Decannulation Stopper, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
78-3132 78-3137 78-3136 78-3138 78-3131 78-3134 78-3135 78-3139
10198-780EA 58.07 USD InStock
10198-780 10198-782 10198-264 10198-784 10194-286 10194-288 10198-778 10193-700
Decannulation Stopper, OR Grade, Sklar
Stoppers PE/LDPE/HDPE Stoppers
Sklar's Decannulation Stopper is a plug used at the beginning of the decannulation process when a patient is no longer relying on the tracheotomy tube to breathe
Unlike a decannulation cap, which is placed over the tracheotomy tube, a stopper fits inside the inner or outer cannula. The outer cannula typically remains inserted when the decannulation process begins, in the event the patient has difficulty breathing through mouth or nose, and requires to begin breathing again through the tube. Latex-free.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty