Would you like to log in again?
Laborde Tracheal Dilator, OR Grade, Sklar
Supplier: Sklar
78-3460
10198-786EA 495.46 USD InStock
10198-786
Laborde Tracheal Dilator, OR Grade, Sklar
Dilators
Sklar Laborde Tracheal Dilator is a non-ratcheted, single spring, finger ring instrument used to widen a tracheal incision to assist in tube insertion and placement.
  • Reusable and latex-free
  • Non sterile
  • Premium OR-grade
When engaged, two outer blades open and a third middle blade pulls in towards the handle. The middle blade is cupped and when closed, the two outer blades rest slightly in the middle blade. This product has straight shanks with three curved, blunt blades.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty