Would you like to log in again?
Kevorkian-Younge Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-6610
10198-864EA 1293.68 USD InStock
10198-864
Kevorkian-Younge Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Kevorkian-Younge Biopsy Punch is a non-ratcheted, finger ring instrument used for obtaining tissue samples in gynecological procedures
The shanks are short and angle at a box lock to a long, narrow shaft that is 10 inches long. The tip has a 3mm x 9.7mm rectangular bite with 4 lower teeth to grasp and excise the tissue for biopsy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty