Would you like to log in again?
Eppendorfer Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-6820
10198-866EA 1442.56 USD InStock
10198-866
Eppendorfer Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Eppendorfer Biopsy Punch is a non-ratcheted, finger ring OB/GYN instrument used for taking cervical tissue samples during procedures such as a colposcopy. Biopsy punches are typically chosen based on user preference, patient size, and the desired size of the sample. The Eppendorfer biopsy punch is considered an anvil style punch. This product has short, straight shanks that angle to a long straight shaft with a 4mm x 8mm biting tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty