Would you like to log in again?
Cannula With Tip for Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
90-7576 90-7579 90-7577 90-7556 90-7558 90-7555 90-7578
10196-400EA 1133.83 USD InStock
10196-400 10198-892 10196-406 10198-886 10196-402 10198-888 10196-404
Cannula With Tip for Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Cannulae
Sklar's Universal Style Biopsy Punch Canula and Tip is part of an OB/GYN instrument
This product attaches to a universal style handle and together the complete instrument they create are biopsy punch forceps. They are often used in colposcopy procedures to obtain cervical tissue samples for biopsy. The bite of this standard Tischler punch is 3mm x 7mm and it produces an oval shaped tissue sample. A standard size biopsy punch may produce greater bleeding than a mini or baby punch; however, with certain diagnoses it is necessary to obtain a larger tissue sample.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty