Would you like to log in again?
Rampley Sponge Forceps, Sklar
Supplier: Sklar
87-2240
10199-042EA 208.44 USD InStock
10199-042
Rampley Sponge Forceps, Sklar
Forceps
The stainless steel construction offers non-corrosive, anti-bacterial and rust-resistant features, providing greater durability.
  • Finger ring handles
  • Serrated fenestrated teardrop jaws
  • Ratchet lock
  • Reusable
  • Latex-free
Sponge forceps primarily used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty