Would you like to log in again?
Sklar Black™ Joseph Skin Hook, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
90-1053 90-1055 90-1051 90-1050 90-1054 90-1052
10199-060EA 353.39 USD InStock
10199-060 10199-058 10199-056 10194-454 10199-064 10199-062
Sklar Black™ Joseph Skin Hook, OR Grade, Sklar
Retractors
The Sklar Black™ joseph skin hook is a handheld retractor commonly used for holding back the edges of skin during laser intranasal and pharyngeal procedures.
  • Safe and durable coating
  • Reduces laser reflection to less than one-tenth of a percent
It is available with a single or dual prong, sharp hook. The handle is often round and knurled and it tapers to the hook end. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.
It ensures a safer surgical environment for patients and surgical staff. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty