Would you like to log in again?
Sklar Black™ Iris Hook, OR Grade, Sklar
Supplier: Sklar
90-2128
10199-074EA 307.3 USD InStock
10199-074
Sklar Black™ Iris Hook, OR Grade, Sklar
Retractors
The Sklar Black™ Iris Hook, is most commonly used in OB/GYN procedures involving the uterus
The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects. This product is straight with a 1mm hook tip and an overall length of 12 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty