Would you like to log in again?
Sklar Black™ Tuttle Forceps, OR Grade, Sklar
Supplier: Sklar
90-2270
10199-094EA 589.44 USD InStock
10199-094
Sklar Black™ Tuttle Forceps, OR Grade, Sklar
Forceps
Sklar Black™ Tuttle Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
These forceps have long shafts which make them useful for working in areas such as the intestines. These forceps may also be known as Singley forceps.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty