Would you like to log in again?
Sklar Black™ Singley (Tuttle) Forceps, OR Grade, Sklar
Supplier: Sklar
90-2272
10199-096EA 616.75 USD InStock
10199-096
Sklar Black™ Singley (Tuttle) Forceps, OR Grade, Sklar
Forceps
Sklar Black™ Singley (Tuttle) Forceps are thumb forceps commonly used for grasping intestinal tissue or dressings materials and sponges during laser procedures.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.
This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. This product is straight with serrated tips and a length of 5 1/2". This producthas oval, fenestrated, serrated tips.
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty