Would you like to log in again?
Heaney Forceps, OR Grade, Sklar
Supplier: Sklar
90-1977 90-2387 90-2280 90-2177
10197-178EA 394.84 USD InStock
10197-178 10199-102 10199-088 10199-130
Heaney Forceps, OR Grade, Sklar
Forceps
Sklar's Heaney Forceps are ratcheted, finger ring forceps used for grasping uterine ligaments during vaginal hysterectomy procedures
They are available straight or curved with one or two teeth. Heaney forceps have diagonal serrations the full length of the jaws, and the jaws are approximately 1/3 the length of the shanks. The combination of serrations and teeth provide a secure hold on the ligaments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty