Would you like to log in again?
Sklar Black™ DeBakey Atraumatic Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
90-2310
10199-114EA 754.34 USD InStock
10199-114
Sklar Black™ DeBakey Atraumatic Tissue Forceps, OR Grade, Sklar
Forceps
Sklar Black™ DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps used for grasping fine tissue during laser surgeries
They are commonly used in vascular and general surgery. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty