Would you like to log in again?
Sklar Black™ Heaney Needle Holder, OR Grade, Sklar
Supplier: Sklar
90-2330
10199-118EA 559.51 USD InStock
10199-118
Sklar Black™ Heaney Needle Holder, OR Grade, Sklar
Forceps
The Sklar Black™ Heaney Needle Holder is a ratcheted, finger ring instrument used for light to medium weight suturing, most commonly in gynecological procedures such as vaginal and abdominal hysterectomy, and dilation and curettage
The slightly curved shanks and tips provide the user a better view when suturing intravaginally, and the cross-serrations provide a secure hold on the needle. This needle holder may also be used as a cardiovascular approximation and closure instrument. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty