Would you like to log in again?
Sklar Black™ Bozeman Forceps, OR Grade, Sklar
Supplier: Sklar
90-2342 90-2340
10199-122EA 446.36 USD InStock
10199-122 10199-120
Sklar Black™ Bozeman Forceps, OR Grade, Sklar
Forceps
Sklar Black™ Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures
They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty