Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Heaney-Ballentine Forceps, OR Grade, Sklar
Supplier: Sklar
90-2480 90-2580
10199-134EA 486.02 USD InStock
10199-134 10199-132
Heaney-Ballentine Forceps, OR Grade, Sklar
Forceps
Sklar's Heaney-Ballentine Forceps are ratcheted, finger ring forceps used for grasping uterine ligaments during vaginal hysterectomy procedures
They are available straight or curved with a single toothed tip. Heaney-Ballentine forceps have longitudinal serrations which distinguish them from standard Heaney forceps, which have diagonal serrations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty