Would you like to log in again?
Quick Release Iris Hook, OR Grade, Sklar
Supplier: Sklar
90-8336 90-8332 90-8344 90-8334 90-8348 90-8335 90-8346
10199-170EA 258.13 USD InStock
10199-170 10199-168 10199-156 10199-166 10199-162 10194-116 10199-160
Quick Release Iris Hook, OR Grade, Sklar
Retractors
The Sklar Iris Hook is a versatile instrument that can be used in many procedures to hold and retract tissue.
It is similar to, but smaller than the Emmett Hook, and the hook is slightly angled. The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty