Would you like to log in again?
Pryor-Pean Retractor, OR Grade, Sklar
Supplier: Sklar
90-3510
10199-234EA 291.89 USD InStock
10199-234
Pryor-Pean Retractor, OR Grade, Sklar
Retractors
Sklar's Pryor-Pean Retractor is a long, handheld OB/GYN retractor
It features a 103/4" long handle that leads to a 1" wide blade. The blade extends 41/2" at a right angle away from the handle. The blade is smooth and rounded at the tip. It has a smooth handle with a hook at the bottom that can function as a finger rest.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty