Would you like to log in again?
Green-Armytage Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
90-5420
10199-290EA 535.15 USD InStock
10199-290
Green-Armytage Hemostatic Forceps, OR Grade, Sklar
Forceps
Green-Armytage Hemostatic Forceps are used to grasp and clamp tissue in the uterus
Censitrac Ready products are industry standard and constructed of stainless steel.
The locking ring handle along with wide blades with grooves gets a firm grip on the tissue. The forceps are available in straight and curved.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty