Would you like to log in again?
Sims Uterine Curette, OR Grade, Sklar
Supplier: Sklar
90-5500 90-5425 90-5424 90-5499 90-5422 90-5426 90-5421 90-5423
10197-990EA 188.04 USD InStock
10197-990 10197-986 10198-006 10199-296 10198-008 10199-294 10197-988 10199-292
Sims Uterine Curette, OR Grade, Sklar
Curettes
Sims Uterine Curette is an instrument used for removing tissue from the lining of the uterus.
  • Hollow with vertical grooves handle
  • Reusable and non sterile
  • Latex-Free
It may be used to scrape and remove growths or tissue to be biopsied. It may also be used as part of a D&C procedure. The teardrop shaped, fenestrated, loop tip is sharp and available in various sizes. The shaft is malleable so that it can be manipulated to an angle suitable for both user and patient needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty