Would you like to log in again?
Adson Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
50-3148 50-3044
10199-334EA 65.56 USD InStock
10199-334 89025-078
Adson Dressing Forceps, OR Grade, Sklar
Forceps
Sklar® Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control.
Sklar® LiteGrip™ instruments feature a lightweight, fenestrated handle
Censitrac Ready products are industry standard and constructed of stainless steel.
They are particularly useful during lengthy procedures when fatigue may arise. The finger holes on the handle also allow grip adjustments as needed by the surgeon.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty