Would you like to log in again?
Joseph Periosteal Elevator, OR Grade, Sklar®
Supplier: Sklar
41-1245 41-1240
10199-404EA 149.04 USD InStock
10199-404 10199-402
Joseph Periosteal Elevator, OR Grade, Sklar®
Retractors
ENT/plastic surgery instrument frequently used in nasal reconstructive procedures.
  • Made of stainless steel
The sharp blade minimizes shredding of the periosteal flap and the curvature allows the surgeon to precisely navigate and elevate the nasal periosteum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty