Would you like to log in again?
DeBakey Bronchus Clamp, OR Grade, Sklar®
Supplier: Sklar
52-2385
10199-410EA 875.05 USD InStock
10199-410
DeBakey Bronchus Clamp, OR Grade, Sklar®
Forceps
Sklar's® DeBakey Atraumatic Bronchus Clamp is a ratcheted, finger ring clamp used primarily in cardiothoracic procedures to clamp the bronchus, the main passageways of the windpipes that take air into the lungs
This product has slightly curved shanks, angled jaws, and slightly curved tips. There are 1x2 rows of fine, atraumatic teeth on the tips and the overall length is 10-1/2 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty