Would you like to log in again?
Potts-Smith Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
52-3270 52-3382 52-3410 52-3310 52-3282
10199-428EA 170.71 USD InStock
10199-428 10199-438 10199-426 10199-436 10199-434
Potts-Smith Dressing Forceps, OR Grade, Sklar
Forceps
Potts-Smith Dressing Forceps are used for changing dressings, and for dissecting fine or single-layer tissue.
Serrated tips provide a solid grip on both materials and tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty