Would you like to log in again?
Stable "Z"® Heated Lid and Controller, Bioptechs Inc.®
Supplier: Bioptechs
40319260812
10199-848KT 1074.58 USD InStock
10199-848
Stable "Z"® Heated Lid and Controller, Bioptechs Inc.®
Controllers Temperature Controllers
Designed for use with the Stable "Z" system to prevent condensation and provide a more uniform temperature gradient across the dish of the Stable "Z".
  • Prevents condensation
  • Prevents outside contamination
Certifications: CE certified.
Delivery information: Includes lid, controller, and power supply.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty