Would you like to log in again?
EDU Culture Dish Control System, Bioptechs Inc.®
Supplier: Bioptechs
0420050421 04200504211 04200504212
10199-856EA 1074.58 USD InStock
10199-856 10199-854 10199-852
EDU Culture Dish Control System, Bioptechs Inc.®
Educational Classroom Kits and Activities
The EDU Culture Dish Control System is a simplified version of the Delta T® system
  • Enhance the instruction of biology
  • Includes curriculum relevant demonstrations
  • Temperature selection options
  • Basic X, Y translation built in
The EDU Starter Set includes all of the necessary components required for full system operation. The EDU Controller and Power Supply is an electronic, closed-loop temperature controller interfaced to EDU stage adapters.
It is designed to facilitate demonstrations of live cellular events in a classroom setting.
Certifications: CE certified.
Ordering information: EDU Starter Set includes, EDU controller, power supply, EDU stage adapter, one pack of 10 dishes, one hundred 22 mm coverslips, 22 mm glass spacer, coverslip weight, and 6 mm culture cylinder. When purchased individually, the controller and power supply comes with a multi-voltage AC input, 12 VDC output power supply. Also available separately, the EDU Stage Adapter is a metal plate containing a circuit board, temperature sensor, and electrical contacts that accept Delta T® dishes and enable X,Y translation of the specimen on any microscope.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty