Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ICD Heated Lid, Bioptechs Inc.®
Supplier: Bioptechs
19030812
10199-872EA 1167.06 USD InStock
10199-872
ICD Heated Lid, Bioptechs Inc.®
Seals Gas Stops
This O-Ring sealed, stainless steel insert is equipped with a heated glass window to prevent condensation and atmospherically seal an ICD.
  • Comes with CO2 port
  • Prevents outside contamination
  • Use with interchangeable coverglass dish
Lid has a gas flow port and allows trans-illumination on an inverted microscope.
Certifications: CE certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty