Would you like to log in again?
FCS2® Open Mode Adapter, Bioptechs Inc.®
Supplier: Bioptechs
06031921513
10200-022EA 468.89 USD InStock
10200-022
FCS2® Open Mode Adapter, Bioptechs Inc.®
Fittings and Connectors
The white, top portion of a FCS2® without perfusion ports, which enables the FCS2® to be assembled without the microaqueduct slide
This enables sealing of the 40 mm coverslip to the base of the chamber. Oftentimes used for microinjection.
Certifications: CE certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty