Would you like to log in again?
Glass Coverslips, 40mm, Bioptechs Inc.®
Supplier: Bioptechs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4013130319 40131303193 40131303192
10200-062PK 943.79 USD InStock
10200-062 10200-060 10200-064
Glass Coverslips, 40mm, Bioptechs Inc.®
Cover Glasses
These 40mm German Desag 263 borosilicate glass coverslips are ideal for imaging applications
  • Ideal for imaging
  • Desag 263 low-alkaline glass for cell adhesion
Available in packages of 200, 500, or 1000.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty