Would you like to log in again?
SP Bel-Art Floating Centrifuge Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F18845-0080 F18845-0050
10204-926CS 140.69 USD InStock
10204-926 10204-928
SP Bel-Art Floating Centrifuge Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Open-style floating rack provides tubes with maximum water contact and better circulation of the water bath around the tubes, allowing superior temperature control of the samples.
  • Floats on water
  • Durable white polypropylene construction
  • Corrosion-resistant
  • Autoclavable
Tubes are held tightly within the rack to assure no "pop-up" tubes or "floaters", while pressing the legs of the rack on the bench will raise tubes and allow for easy removal. Racks are made of durable, corrosion-resistant white polypropylene plastic. They feature end handles to allow easy transport and stacking. Steam autoclavable at 121°C (250°F).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty