Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Troponin T Mouse Monoclonal Antibody [clone: TT-98]

Supplier: Boster
MA1098
10205-980EA 809.04 USD InStock
10205-980
Anti-Troponin T Mouse Monoclonal Antibody [clone: TT-98]
Antibodies
Mouse IgG monoclonal antibody for Troponin T, troponin T type 1 (skeletal, slow); troponin T type 2 (cardiac); troponin T type 3 (skeletal, fast) (TNNT1; TNNT2; TNNT3) detection. Tested with WB, IHC-P, IHC-F in Human;mouse;rat;rabbit;chicken. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Troponin T
Clonality: monoclonal
Clone: TT-98
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat, Rabbit, Chicken

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty