Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GCG Mouse Monoclonal Antibody [clone: IMD-7]

Supplier: Boster
MA1047
10206-024EA 808.82 USD InStock
10206-024
Anti-GCG Mouse Monoclonal Antibody [clone: IMD-7]
Antibodies
Mouse IgG monoclonal antibody for Glucagon, glucagon (GCG) detection. Tested with IHC-P in Human, mouse, rat, rabbit. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Glucagon
Clonality: monoclonal
Clone: IMD-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty