Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFAP Polyclonal Antibody

Supplier: Boster
PB9082
10207-464EA 808.99 USD InStock
10207-464
Anti-GFAP Polyclonal Antibody
Antibodies
Polyclonal antibody for GFAP detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. GFAP information: Molecular Weight: 49880 MW; Subcellular Localization: Cytoplasm . Associated with intermediate filaments; Tissue Specificity: Expressed in cells lacking fibronectin.

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty