Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFSF4 Rat Monoclonal Antibody (Biotin) [clone: RM134L]

Supplier: Tonbo Biosciences
30-5905-U025 30-5905-U100
10208-344EA 38.4 USD InStock
10208-344 10208-346
Anti-TNFSF4 Rat Monoclonal Antibody (Biotin) [clone: RM134L]
Antibodies
The RM134L antibody recognizes CD252, also known as OX-40 Ligand or CD134 Ligand, a member of the TNF superfamily that is present on the surface of antigen presenting cells and activated B lymphocytes. The OX-40 Ligand interacts with OX-40 (CD134) which is expressed primarily on activated T cells. This costimulatory interaction leads to increased proliferation and IL-2 production responses of activated T cells, and at the same time enhances proliferation and immunoglobulin secretion by activated B cells.

The RM134L antibody is useful for flow cytometric detection of CD252 on stimulated mouse splenocytes. It has also been reported to block the costimulatory activity of OX-40 Ligand. Please choose the appropriate format for each application.

Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: RM134L
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty