Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 61D3]

Supplier: Tonbo Biosciences
35-0149-T025 35-0149-T100
10208-364EA 63.6 USD InStock
10208-364 10208-366
Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 61D3]
Antibodies
The 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty