Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL10 Rat Monoclonal Antibody [clone: JES5-2A5]

Supplier: Tonbo Biosciences
40-7102-U100 40-7102-U500
10208-430EA 215 USD InStock
10208-430 10208-428
Anti-IL10 Rat Monoclonal Antibody [clone: JES5-2A5]
Antibodies
The JES5-2A5 antibody is specific for mouse IL-10. IL-10 has numerous biological activities including the ability to inhibit both the synthesis of Th1 cytokines and proliferation of T cells. In the mouse, IL-10 is produced by phagocytic cells, conventional dendritic, cells, T cells, B cells, and NK cells. The JES5-2A5 antibody is a neutralizing antibody.

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: JES5-2A5
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty