Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD8A Mouse Monoclonal Antibody (violetFluor® 450) [clone: Hit8a]

Supplier: Tonbo Biosciences
75-0089-T100 75-0089-T025
10208-494EA 174 USD InStock
10208-494 10208-492
Anti-CD8A Mouse Monoclonal Antibody (violetFluor® 450) [clone: Hit8a]
Antibodies
The Hit8a antibody is specific for the 32-34 kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.

The Hit8a antibody is widely used as a phenotypic marker for CD8 on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells. If used together with an alternative Anti-Human CD8a clone RPA-T8, the Hit8a antibody will not block binding of RPA-T8 to CD8a.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: Hit8a
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty