Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAX Mouse Monoclonal Antibody (violetFluor® 450) [clone: 3.9]

Supplier: Tonbo Biosciences
75-0116-T025 75-0116-T100
10208-496EA 88.8 USD InStock
10208-496 10208-498
Anti-ITGAX Mouse Monoclonal Antibody (violetFluor® 450) [clone: 3.9]
Antibodies
The 3.9 antibody reacts with human CD11c, also known as integrin alpha X. This 150 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha X (CD11c) assembles with Integrin beta-2 (CD18) into a receptor known as CR4 which can bind and induce signaling through ICAMs and VCAM-1 on endothelial cells and can also facilitate removal of iC3b bearing foreign cells.

The 3.9 antibody is widely used as a marker for CD11c expression on dendritic cells (DC), often in parallel with markers for CD11b, for identification of developmental stages and mature subsets of this cell type. CD11c is prominently expressed on tissue macrophages, and is also detected on activated neutrophils, granulocytes, some types of activated T cells and intestinal intraepithelial lymphocytes (IEL). The antibody is reported to be cross-reactive with Baboon, Chimpanzee, Cynomolgus and Rhesus CD11c.

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 3.9
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Rhesus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty