Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Progesterone Receptor Polyclonal Antibody

Supplier: Boster
PB9009
10209-130EA 808.99 USD InStock
10209-130
Anti-Progesterone Receptor Polyclonal Antibody
Antibodies
Polyclonal antibody for NR3C3/PGR detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. NR3C3/PGR information: Molecular Weight: 98981 MW; Subcellular Localization: Nucleus. Cytoplasm. Nucleoplasmic shuttling is both homone- and cell cycle-dependent. On hormone stimulation, retained in the cytoplasm in the G(1) and G(2)/M phases.

Type: Primary
Antigen: Progesterone Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty