Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cathepsin B Polyclonal Antibody

Supplier: Boster
PB9046
10209-222EA 808.99 USD InStock
10209-222
Anti-Cathepsin B Polyclonal Antibody
Antibodies
Polyclonal antibody for Cathepsin B/CTSB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Cathepsin B/CTSB information: Molecular Weight: 37822 MW; Subcellular Localization: Lysosome. Melanosome. Secreted, extracellular space . Identified by mass spectrometry in melanosome fractions from stage I to stage IV.

Type: Primary
Antigen: Cathepsin B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty