Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cyclin D1 Polyclonal Antibody

Supplier: Boster
PA1245-2
10209-310EA 809.04 USD InStock
10209-310
Anti-Cyclin D1 Polyclonal Antibody
Antibodies
Polyclonal antibody for Cyclin D1/CCND1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Cyclin D1/CCND1 information: Molecular Weight: 33729 MW; Subcellular Localization: Nucleus . Cytoplasm . Membrane . Cyclin D-CDK4 complexes accumulate at the nuclear membrane and are then translocated to the nucleus through interaction with KIP/CIP family members.

Type: Primary
Antigen: Cyclin D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty