Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Hamartin Polyclonal Antibody

Supplier: Boster
PB9120
10209-434EA 808.99 USD InStock
10209-434
Anti-Hamartin Polyclonal Antibody
Antibodies
Polyclonal antibody for HAMARTIN/TSC1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. HAMARTIN/TSC1 information: Molecular Weight: 129767 MW; Subcellular Localization: Cytoplasm . Membrane ; Peripheral membrane protein . At steady state found in association with membranes; Tissue Specificity: Highly expressed in skeletal muscle, followed by heart, brain, placenta, pancreas, lung, liver and kidney. Also expressed in embryonic kidney cells.

Type: Primary
Antigen: Hamartin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty