Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Tuberin Polyclonal Antibody

Supplier: Boster
PB9121
10209-436EA 808.99 USD InStock
10209-436
Anti-Tuberin Polyclonal Antibody
Antibodies
Polyclonal antibody for P TUBERIN/TSC2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. P TUBERIN/TSC2 information: Molecular Weight: 200608 MW; Subcellular Localization: Cytoplasm. Membrane; Peripheral membrane protein. At steady state found in association with membranes; Tissue Specificity: Liver, brain, heart, lymphocytes, fibroblasts, biliary epithelium, pancreas, skeletal muscle, kidney, lung and placenta.

Type: Primary
Antigen: Tuberin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty