Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-complement C5a Polyclonal Antibody

Supplier: Boster
PA2308
10209-502EA 808.82 USD InStock
10209-502
Anti-complement C5a Polyclonal Antibody
Antibodies
Polyclonal antibody for C5A/C5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Mouse. C5A/C5 information: Molecular Weight: 8981 MW; Subcellular Localization: Secreted.

Type: Primary
Antigen: complement C5a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty