Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD20 Polyclonal Antibody

Supplier: Boster
PB9050
10209-700EA 808.99 USD InStock
10209-700
Anti-CD20 Polyclonal Antibody
Antibodies
Polyclonal antibody for CD20/MS4A1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. CD20/MS4A1 information: Molecular Weight: 33077 MW; Subcellular Localization: Cell membrane ; Multi-pass membrane protein . Cell membrane ; Lipid-anchor ; Tissue Specificity: Expressed on B-cells.

Type: Primary
Antigen: CD20
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty