Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD23/FCER2 Polyclonal Antibody

Supplier: Boster
PB9051
10209-864EA 808.99 USD InStock
10209-864
Anti-CD23/FCER2 Polyclonal Antibody
Antibodies
Polyclonal antibody for CD23/FCER2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CD23/FCER2 information: Molecular Weight: 37648 MW; Subcellular Localization: Cell membrane ; Single-pass type II membrane protein . Cell membrane ; Lipid-anchor .

Type: Primary
Antigen: CD23/FCER2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty