Would you like to log in again?
Mouse VEGFR3/FLT4 PicoKine ELISA Kit, Boster
Supplier: Boster
PicoKine
EK0546
10209-892EA 947.69 USD InStock
10209-892
Mouse VEGFR3/FLT4 PicoKine ELISA Kit, Boster
Assays ELISAs
Sandwich ELISA kit of Quantitative Detection for Mouse VEGFR3/FLT4
Ordering information: Kits include 96-well plate precoated with anti- mouse VEGFR3 antibody 1; ST0000-10 Lyophilized mouse VEGFR3 standard 10ng/tubex2; AR1107 Biotinylated anti- mouse VEGFR3 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty