Would you like to log in again?
AMF Active Air Self Leveling Platform, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6410 6280 6350 6310 6400 6270 6380 6300 6320 6360 6370 6260 6250 6340 6390 6290 6330 6200
102090-818EA 5474.32 USD InStock
102090-818 102090-828 102090-838 102090-826 102090-848 102090-836 102090-824 102090-846 102090-834 102090-844 102090-822 102090-854 102090-832 102090-842 102090-820 102090-830 102090-840 102090-850
AMF Active Air Self Leveling Platform, Electron Microscopy Sciences
Support Stands
Platforms and workstations improve the environment for microscopes with all the features necessary to make your work easier
To select a platform you must first decide on the size of the working surface you require, the load it needs to support and the sensitivity of the equipment to vibrations; from this you can determine the class of product you require. You then need to decide whether you will require it to be floor mounted and what type of air supply would be best suited. These platforms are self-levelling and ideal for use when loads on the platform are likely to change. They can support up to 160kg and are available in a range of standard sizes from 60x60cm up to 150x90cm. Their low 6cm working height and low weight makes them ideal for use on desks and benches. The AMF and AMP platforms offer the ultimate performance, incorporating four 2Hz damped rolling diaphragm air isolators, efficiently isolating frequencies from 6Hz and upwards. Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilisation times and speeding up measurements. These platforms are designed to be pumped-up and left alone. Simple to use, they are ideal for use with fixed loads or in applications where pressurised air or gas cannot be used. They support loads up to 160kg and sizes range from 60 x 60 cm to 150 x 90 cm with a 6cm working height.
Workstations provide a tailored working environment as well as featuring an isolation platform built into the work surface, making working with a microscope both faster and easier, leading to more reliable measurements in your laboratory. Platforms can also be used stand alone beneath microscopes to isolate them from external disturbances and speed up measurement. The AMF and AMP platforms offer the ultimate performance, incorporating four 2Hz damped rolling diaphragm air isolators, efficiently isolating frequencies from 6Hz and upwards. The AMS tables incorporate Sorbothane isolators, giving excellent isolation with a natural frequency of less than 7Hz. Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilisation times and speeding up measurements. The ergonomic workstation design minimises user stress. With the active area option microscope platforms sit flush with the work surface and the surrounding area is useable for general work without disturbing measurements. Frames have height adjustable levelling feet, for excellent stability and user comfort with retractable castor feet, frame shelf and monitor support optional.
Ordering information: Available with standard white epoxy or optional Stainless, Ferromagnetic, or Stainless 316 steel top.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty