Would you like to log in again?
Agarose, White, Fine Powder and Tablets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
10206 10205
102090-958PK 305.41 USD InStock
102090-958 100503-642
Agarose, White, Fine Powder and Tablets, Electron Microscopy Sciences
Electrophoresis Gels Agarose and Acrylamide
Agarose I™ is standard melting/gelling agarose, suitable for routine nucleic acid analytical/preservative applications
  • CAS No.= 9012-36-6
Agarose I™ has a low EEO for shorter electrophoresis runs without compromising resolution. Available in fine powder or tablet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty