Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Anti-IgG Goat Polyclonal Antibody (Gold, >1 nm (ultra-small))

Supplier: Electron Microscopy Sciences
25900 25901
102091-026EA 766.62 USD InStock
102091-026 102091-028
Anti-IgG Goat Polyclonal Antibody (Gold, >1 nm (ultra-small))
Antibodies
These antibodies can be used for double labeling as well when both mouse and rat primary antibodies are used with other than mouse or rat tissue. Cross adsorbed against Rat immunoglobulins.

Cross reactivity can be a problem when working with primary antibodies sourced from animal species that are evolutionary closely related to the source of tissues or cells that antigens are to be detected in.These antibodies can be used for double labeling as well when both mouse and rat primary antibodies are used with other than mouse or rat tissue. Cross adsorbed against Rat immunoglobulins.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, >1 nm (ultra-small)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty