Would you like to log in again?
Tissue Embedding Center, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63120
102091-084EA 18405.35 USD InStock
102091-084
Tissue Embedding Center, Electron Microscopy Sciences
Tissue Embedders
The Histo-Pro 150 offers superior operating convenience for tissue embedding processes in a modular ergonomic design.
  • Two Module system (Histo Pro 150H and Histo Pro 150CS)
  • Fully programmable for both time and temperature
System includes: Four liter paraffin tank, self draining forceps holder, auto defrosting cold plate, a large cold plate, cassette bath and a user friendly proprietary PAT interface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty